This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAP3K19 SIRNA Assays ›
Gene Symbol:
MAP3K19
Gene Name:
mitogen-activated protein kinase kinase kinase 19
Gene Aliases:
RCK, YSK4
Chromosome Location:
Chr.2: 134963875 - 135047468 on Build GRCh38
UniGene ID:
Hs.659395
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188353
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAP3K19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018044.2 NP_001018054.1 7 2970
NM_001018046.2 NP_001018056.1 8 855
NM_001282883.1 NP_001269812.1 7 705
NM_001321177.1 NP_001308106.1 10 3601
NM_025052.4 NP_079328.3 9 3309
XM_011511891.2 XP_011510193.1 9 3449
XM_011511892.1 XP_011510194.1 12 4017
XM_011511894.2 XP_011510196.1 9 3424
XM_011511896.2 XP_011510198.1 9 3290
XM_011511897.2 XP_011510199.1 9 3370
XM_017005003.1 XP_016860492.1 12 3984
XM_017005004.1 XP_016860493.1 11 3869
XM_017005005.1 XP_016860494.1 10 3670
XM_017005006.1 XP_016860495.1 10 3032
XM_017005007.1 XP_016860496.1 9 3417
GenBank mRNA AK026727.1 - 1 190
AK122935.1 - 10 3601
AK131216.1 - 3 1448
AK225579.1 - 1 190
AY574901.1 - 9 3309
AY574902.1 - 7 2970
AY574903.1 - 9 3309
AY574904.1 - 8 855
BC034417.1 - 2 356
BC137276.1 - 9 3308 9021273
BC137277.1 - 9 3308 9021274
BC144555.1 - 7 704

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction by protein phosphorylation
actin cytoskeleton organization
receptor signaling protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0