This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF703 SIRNA Assays ›
Gene Symbol:
ZNF703
Gene Name:
zinc finger protein 703
Gene Aliases:
NLZ1, ZEPPO1, ZNF503L, ZPO1
Chromosome Location:
Chr.8: 37695751 - 37700021 on Build GRCh38
UniGene ID:
Hs.288042
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188359
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF703
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025069.2 NP_079345.1 1 379
GenBank mRNA AK024361.1 - 1 347
BC032534.1 - 1 308 5527569
BC084581.1 - 1 341
DA936465.1 - 1 381

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation
positive regulation of epithelial to mesenchymal transition
regulation of transforming growth factor beta receptor signaling pathway
positive regulation of cell migration
positive regulation of mammary gland epithelial cell proliferation
negative regulation of homotypic cell-cell adhesion
adherens junction assembly
negative regulation of transcription, DNA-templated
regulation of cell cycle
mammary gland epithelial cell differentiation
regulation of canonical Wnt signaling pathway
cellular response to estradiol stimulus
nucleic acid binding
protein binding
metal ion binding
repressing transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline