siRNA Details
| Gene Symbol: UBXN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171091.1 | NP_001164562.1 | 2 | 152 | ||
| NM_025241.2 | NP_079517.1 | 2 | 254 | |||
| XM_017027325.1 | XP_016882814.1 | 3 | 685 | |||
| GenBank mRNA | AF272893.1 | - | 2 | 253 | ||
| AF272894.1 | - | 2 | 143 | |||
| BC008288.1 | - | 2 | 152 | 3659797 | ||
| BM764980.1 | - | 2 | 254 | |||

UBXN6
Genomic Map