siRNA Details
| Gene Symbol: FBXO38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271723.1 | NP_001258652.1 | 8 | 1066 | ||
| NM_030793.4 | NP_110420.3 | 8 | 1066 | |||
| XM_005268513.1 | XP_005268570.1 | 8 | 1055 | |||
| XM_006714797.1 | XP_006714860.1 | 8 | 1055 | |||
| XM_011537684.2 | XP_011535986.1 | 7 | 961 | |||
| XM_017009899.1 | XP_016865388.1 | 8 | 1044 | |||
| XM_017009900.1 | XP_016865389.1 | 7 | 961 | |||
| XM_017009901.1 | XP_016865390.1 | 7 | 961 | |||
| XM_017009902.1 | XP_016865391.1 | 7 | 961 | |||
| GenBank mRNA | AK024024.1 | - | 8 | 986 | ||
| BC033454.1 | - | 8 | 999 | 4510274 | ||
| BC050424.1 | - | 7 | 971 | 6472693 | ||

FBXO38
Genomic Map