This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GFOD2 SIRNA Assays ›
Gene Symbol:
GFOD2
Gene Name:
glucose-fructose oxidoreductase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.16: 67674533 - 67719370 on Build GRCh38
UniGene ID:
Hs.307084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS188600
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GFOD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030819.3 NP_110446.3 3 646
NR_027398.1 2 300
XM_006721288.3 XP_006721351.1 3 389
XM_017023738.1 XP_016879227.1 3 596
GenBank mRNA AK074382.1 - 4 599
AK091606.1 - 1 1523
AK225997.1 - 3 616
AL832904.1 - 1 85
AY358169.1 - 2 356
BC000757.2 - 2 253 2820364
BC002943.1 - 2 273
BC107585.1 - 3 596 6463662
BI826603.1 - 3 700

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

extracellular matrix organization
oxidation-reduction process
oxidoreductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline