siRNA Details
| Gene Symbol: ZDHHC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032283.2 | NP_115659.1 | 8 | 1234 | ||
| GenBank mRNA | AK056427.1 | - | 8 | 986 | ||
| AK298683.1 | - | 8 | 841 | |||
| AL512765.1 | - | 8 | 839 | |||
| BC066776.1 | - | 8 | 1234 | 30342061 | ||

ZDHHC18
Genomic Map