This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM185A SIRNA Assays ›
Gene Symbol:
TMEM185A
Gene Name:
transmembrane protein 185A
Gene Aliases:
CXorf13, FAM11A, FRAXF, ee3
Chromosome Location:
Chr.X: 149596556 - 149631912 on Build GRCh38
UniGene ID:
Hs.522172
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS189107
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM185A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282302.1 NP_001269231.1 2 475
NM_032508.3 NP_115897.1 2 475
GenBank mRNA AF530473.1 - 3 454
AK096308.1 - 2 287
BC103820.1 - 2 179
BC103821.1 - 2 179 40000939
BC103822.1 - 2 179 40000940
BG721461.1 - 1 74
BQ224383.1 - 2 266
DA206736.1 - 2 367

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-cmcqw:80/100.66.72.176:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0