This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNHIT3 SIRNA Assays ›
Gene Symbol:
ZNHIT3
Gene Name:
zinc finger HIT-type containing 3
Gene Aliases:
TRIP3
Chromosome Location:
Chr.17: 36486627 - 36499310 on Build GRCh38
UniGene ID:
Hs.2210
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS190196
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNHIT3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281432.1 NP_001268361.1 4 336
NM_001281433.1 NP_001268362.1 4 302
NM_004773.3 NP_004764.1 4 336
NR_104009.1 4 331
NR_104010.1 3 304
NR_104011.1 3 304
GenBank mRNA AB209193.1 - 2 3800
AF400652.1 - 4 328
AK290858.1 - 4 287
AK308583.1 - 3 232
AV763423.1 - 299
BC017931.1 - 4 301 4669658
BC105608.1 - 3 276
BG032116.1 - 4 237
BI767446.1 - 4 269
BM457955.1 - 266
BQ009986.1 - 2 635
HY045632.1 - 4 337
L40410.1 - 4 254

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
protein binding
metal ion binding
thyroid hormone receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline