This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RCC2 SIRNA Assays ›
Gene Symbol:
Rcc2
Gene Name:
regulator of chromosome condensation 2
Gene Aliases:
2610510H01Rik, 2610529N02Rik, AA536646, AA675016, Td60, mKIAA1470
Chromosome Location:
Chr.4: 140701473 - 140723220 on Build GRCm38
UniGene ID:
Mm.253
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS201075
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rcc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173867.5 NP_776292.1 7 989
GenBank mRNA AK076040.1 - 7 920
AK076046.1 - 1220
AK129370.1 - 6 693
AK204705.1 - 1 25
BC020385.1 - 5 522
BC034759.1 - 4 308
BC060624.1 - 7 943
BC086666.1 - 7 912 30634817

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
endosome organization
cell cycle
mitotic nuclear division
integrin-mediated signaling pathway
regulation of fibroblast migration
positive regulation of G2/M transition of mitotic cell cycle
regulation of cell migration
negative regulation of GTPase activity
establishment of protein localization
focal adhesion assembly
cell division
negative regulation of focal adhesion assembly
positive regulation of attachment of spindle microtubules to kinetochore
chromosome passenger complex localization to kinetochore
activation of GTPase activity
negative regulation of substrate adhesion-dependent cell spreading
regulation of ruffle assembly
microtubule binding
protein kinase binding
protein domain specific binding
small GTPase binding
poly(A) RNA binding
Rac GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline