siRNA Details
| Gene Symbol: Chd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177224.2 | NP_796198.1 | 3 | 995 | ||
| GenBank mRNA | AK034446.1 | - | 3 | 996 | ||
| AK039562.1 | - | 3 | 995 | |||
| AK084000.1 | - | 2 | 982 | |||
| BC145219.1 | - | 1 | 694 | 9053770 | ||
| BC145220.1 | - | 1 | 694 | |||
| DQ127229.2 | - | 3 | 995 | |||

Chd9
Genomic Map