siRNA Details
| Gene Symbol: Chd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177224.2 | NP_796198.1 | 4 | 2000 | ||
| GenBank mRNA | AK030430.1 | - | 2 | 464 | ||
| AK034446.1 | - | 4 | 2001 | |||
| AK039562.1 | - | 4 | 2000 | |||
| AK040994.1 | - | 2 | 429 | |||
| AK084000.1 | - | 3 | 1987 | |||
| DQ127229.2 | - | 4 | 2000 | |||

Chd9
Genomic Map