This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HSPA13 SIRNA Assays ›
Gene Symbol:
Hspa13
Gene Name:
heat shock protein 70 family, member 13
Gene Aliases:
1600002I10Rik, AV006182, B230217N24Rik, Stch
Chromosome Location:
Chr.16: 75755190 - 75767277 on Build GRCm38
UniGene ID:
Mm.26704
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS201441
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hspa13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030201.3 NP_084477.1 3 441
GenBank mRNA AK021006.1 - 3 413
AK030553.1 - 3 414
AK034643.1 - 3 421
AK049011.1 - 3 409
AK167839.1 - 3 384
BC039701.1 - 2 382
BC085181.1 - 3 401 30646642
BC094508.1 - 3 384

Back To Top

More Information


Gene Ontology Categories:

Process(es)

nucleotide binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline