siRNA Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017366.3 | NP_059062.1 | 16, 17 | 1721 | ||
| GenBank mRNA | AF017176.1 | - | 13, 14 | 1369 | ||
| AK076037.1 | - | 16, 17 | 1682 | |||
| AK146334.1 | - | 11, 12 | 1194 | |||
| BC026559.1 | - | 16, 17 | 1700 | 5037878 | ||
| U41497.1 | - | 7, 8 | 603 | |||
| Z71189.1 | - | 16, 17 | 1642 | |||

Acadvl
Genomic Map