siRNA Details
| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013463.2 | NP_038491.2 | 3, 4 | 625 | ||
| GenBank mRNA | AK040081.1 | - | 3, 4 | 557 | ||
| AK054547.1 | - | 3, 4 | 577 | |||
| AK145011.1 | - | 3, 4 | 567 | |||
| AK145148.1 | - | 625 | ||||
| AK164231.1 | - | 3, 4 | 558 | |||
| AK166204.1 | - | 3, 4 | 558 | |||
| AK166245.1 | - | 3, 4 | 574 | |||
| BC009021.1 | - | 3, 4 | 578 | 3708950 | ||
| U34071.1 | - | 3, 4 | 575 | |||
| U50716.1 | - | 3, 4 | 563 | |||

Gla
Genomic Map