siRNA Details
| Gene Symbol: Ptk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130409.1 | NP_001123881.1 | 10, 11 | 1258 | ||
| NM_007982.2 | NP_032008.2 | 9, 10 | 1103 | |||
| GenBank mRNA | AB011499.1 | - | 7, 8 | 791 | ||
| AB030035.1 | - | 7, 8 | 791 | |||
| AK079821.1 | - | 10, 11 | 1355 | |||
| AK220543.1 | - | 9, 10 | 1103 | |||
| BC030180.1 | - | 10, 11 | 1188 | |||
| JN971016.1 | - | 7, 8 | 791 | |||
| M95408.1 | - | 8, 9 | 900 | |||

Ptk2
Genomic Map