siRNA Details
| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008212.4 | NP_032238.2 | 2 | 273 | ||
| GenBank mRNA | AK075609.1 | - | 2 | 200 | ||
| AK132260.1 | - | 2 | 236 | |||
| AK148481.1 | - | 2 | 197 | |||
| AK148486.1 | - | 2 | 210 | |||
| AK167024.1 | - | 2 | 200 | |||
| AK168238.1 | - | 2 | 220 | |||
| AK168877.1 | - | 2 | 200 | |||
| AK169261.1 | - | 2 | 239 | |||
| BC028833.1 | - | 2 | 105 | |||
| BC064712.1 | - | 2 | 215 | 30303218 | ||
| D29639.1 | - | 2 | 199 | |||

Hadh
Genomic Map