siRNA Details
| Gene Symbol: Cep250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129999.1 | NP_001123471.1 | 28 | 5632 | ||
| NM_001130000.1 | NP_001123472.1 | 28 | 5630 | |||
| NM_008383.3 | NP_032409.3 | 28 | 5572 | |||
| NM_177217.3 | NP_796191.2 | 28 | 5567 | |||
| GenBank mRNA | BC068153.1 | - | 26 | 4809 | ||
| BC075730.1 | - | 1 | 258 | |||
| BC086623.1 | - | 10 | 4014 | |||
| BC145268.1 | - | 25 | 5212 | |||
| BC150746.1 | - | 27 | 5394 | 9087657 | ||
| DQ148475.1 | - | 27 | 5367 | |||

Cep250
Genomic Map