This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KCNQ1 SIRNA Assays ›
Gene Symbol:
Kcnq1
Gene Name:
potassium voltage-gated channel, subfamily Q, member 1
Gene Aliases:
AW559127, KVLQT1, Kcna9
Chromosome Location:
Chr.7: 143106742 - 143427050 on Build GRCm38
UniGene ID:
Mm.439769
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS205730
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnq1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008434.2 NP_032460.2 4 721
GenBank mRNA AB079603.1 - 4 424
AK154244.1 - 4 695
AY331142.1 - 4 636
BC045142.1 - 4 721 6314642
BC055304.1 - 4 676 6397978
U70068.1 - 4 527

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_032073, U92655, AJ133685, AY043394.

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
regulation of gene expression by genetic imprinting
transport
ion transport
potassium ion transport
positive regulation of heart rate
gene silencing
regulation of ion transmembrane transport
cellular response to drug
regulation of membrane potential
negative regulation of insulin secretion
transmembrane transport
cardiac muscle contraction
regulation of membrane repolarization
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of atrial cardiac muscle cell membrane repolarization
positive regulation of cardiac muscle contraction
positive regulation of gastric acid secretion
cellular response to cAMP
potassium ion export
potassium ion transmembrane transport
response to anesthetic
cardiovascular system development
ventricular cardiac muscle cell action potential
membrane repolarization during action potential
membrane repolarization during cardiac muscle cell action potential
atrial cardiac muscle cell action potential
regulation of heart rate by cardiac conduction
potassium ion export across plasma membrane
positive regulation of potassium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
potassium channel activity
calmodulin binding
protein phosphatase 1 binding
outward rectifier potassium channel activity
protein kinase A catalytic subunit binding
protein kinase A regulatory subunit binding
protein homodimerization activity
ion channel binding
voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
voltage-gated potassium channel activity involved in atrial cardiac muscle cell action potential repolarization
scaffold protein binding
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jrd88:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0