siRNA Details
| Gene Symbol: Lamb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008483.3 | NP_032509.2 | 29 | 4706 | ||
| GenBank mRNA | AK177474.1 | - | 2 | 164 | ||
| AK219498.1 | - | 2 | 164 | |||
| BC026051.1 | - | 29 | 4633 | 4036357 | ||
| CK634059.1 | - | 2 | 122 | |||

Lamb2
Genomic Map