siRNA Details
| Gene Symbol: Plcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013829.2 | NP_038857.1 | 18, 19 | 1634 | ||
| GenBank mRNA | AF332071.1 | - | 18, 19 | 1634 | ||
| AF332072.1 | - | 18, 19 | 1634 | |||
| AK133360.1 | - | 1437 | ||||
| BC089521.1 | - | 18, 19 | 1597 | |||
| BC129883.1 | - | 18, 19 | 1629 | 40110646 | ||

Plcb4
Genomic Map