This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRL7A1 SIRNA Assays ›
Gene Symbol:
Prl7a1
Gene Name:
prolactin family 7, subfamily a, member 1
Gene Aliases:
AA407021, AA409971, AA410116, PLP-E, PLP-G, Prlpe, Prlpg
Chromosome Location:
Chr.13: 27633364 - 27642493 on Build GRCm38
UniGene ID:
Mm.196424
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS207988
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prl7a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164058.1 NP_001157530.1 5 358
NM_008930.2 NP_032956.1 5 355
GenBank mRNA AF011381.1 - 5 347
AF020525.1 - 5 329
AK028371.1 - 5 356
AK141309.1 - 5 359
AK146050.1 - 5 359
BB611867.1 - 5 356
BC051671.1 - 5 340 30018275

Back To Top

More Information


Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
hormone activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fxmhk:80/100.66.79.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0