This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PTPN22 SIRNA Assays ›
Gene Symbol:
Ptpn22
Gene Name:
protein tyrosine phosphatase, non-receptor type 22 (lymphoid)
Gene Aliases:
70zpep, PEP, Ptpn8
Chromosome Location:
Chr.3: 103856804 - 103912252 on Build GRCm38
UniGene ID:
Mm.395
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS208234
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptpn22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008979.2 NP_033005.1 2, 3 331
GenBank mRNA AK153812.1 - 4, 5 357
AK169559.1 - 2, 3 288
BC055377.1 - 2, 3 253 30077916
BY092932.1 - 2, 3 333
M90388.1 - 2, 3 316

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106460, XM_006233072, XM_008761379.

Gene Ontology Categories:

Function(s) Process(es)

immune system process
protein dephosphorylation
autophagy
dephosphorylation
T cell differentiation
regulation of natural killer cell proliferation
peptidyl-tyrosine dephosphorylation
phosphoanandamide dephosphorylation
T cell receptor signaling pathway
regulation of T cell receptor signaling pathway
negative regulation of T cell receptor signaling pathway
negative regulation of T cell activation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
hydrolase activity
phosphatase activity
SH3 domain binding
kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0