siRNA Details
| Gene Symbol: Igdcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008988.2 | NP_033014.1 | 11, 12 | 2115 | ||
| GenBank mRNA | AF026465.1 | - | 11, 12 | 2115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igdcc3| Gene Symbol: Igdcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008988.2 | NP_033014.1 | 11, 12 | 2115 | ||
| GenBank mRNA | AF026465.1 | - | 11, 12 | 2115 | ||