siRNA Details
| Gene Symbol: Scn10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205321.1 | NP_001192250.1 | 23 | 4100 | ||
| NM_009134.3 | NP_033160.2 | 23 | 4097 | |||
| GenBank mRNA | AK157866.1 | - | 23 | 4097 | ||
| AY538273.1 | - | 22 | 4027 | |||
| Y09108.1 | - | 22 | 4065 | |||

Scn10a
Genomic Map