siRNA Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009261.3 | NP_033287.2 | 5 | 762 | ||
| GenBank mRNA | BC014710.1 | - | 6 | 846 | 4502840 | |
| X84692.1 | - | 699 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Strbp| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009261.3 | NP_033287.2 | 5 | 762 | ||
| GenBank mRNA | BC014710.1 | - | 6 | 846 | 4502840 | |
| X84692.1 | - | 699 | ||||