siRNA Details
| Gene Symbol: Camkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199676.1 | NP_001186605.1 | 13 | 1521 | ||
| NM_145358.2 | NP_663333.1 | 13 | 1521 | |||
| GenBank mRNA | AF453383.1 | - | 12 | 1304 | ||
| AK031399.1 | - | 13 | 1446 | |||
| AK032070.1 | - | 13 | 1494 | |||
| AK044660.1 | - | 13 | 1510 | |||
| AK122370.1 | - | 8 | 687 | |||
| BC023103.1 | - | 12 | 1354 | 3993474 | ||
| BC025895.1 | - | 2 | 104 | |||
| CK625345.1 | - | 1 | 51 | |||

Camkk2
Genomic Map