siRNA Details
| Gene Symbol: Stat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009284.2 | NP_033310.2 | 14, 15 | 1902 | ||
| GenBank mRNA | AF481808.1 | - | 13, 14 | 1617 | ||
| AF481809.1 | - | 13, 14 | 1617 | |||
| AK149563.1 | - | 14, 15 | 1862 | |||
| AK163053.1 | - | 14, 15 | 1903 | |||
| BC029318.1 | - | 1, 2 | 91 | |||
| BC127054.1 | - | 14, 15 | 1692 | 40109712 | ||

Stat6
Genomic Map