siRNA Details
| Gene Symbol: Depdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144804.1 | NP_659053.1 | 2 | 386 | ||
| GenBank mRNA | AK134830.1 | - | 2 | 406 | ||
| AK144066.1 | - | 2 | 399 | |||
| AK162086.1 | - | 2 | 409 | |||
| AK162218.1 | - | 2 | 409 | |||
| AK165504.1 | - | 2 | 399 | |||
| BC013499.1 | - | 2 | 386 | 4222591 | ||

Depdc7
Genomic Map