This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 4930486L24RIK SIRNA Assays ›
Gene Symbol:
4930486L24Rik
Gene Name:
RIKEN cDNA 4930486L24 gene
Gene Aliases:
AW545031
Chromosome Location:
Chr.13: 60842612 - 60864416 on Build GRCm38
UniGene ID:
Mm.19839
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS210804
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930486L24Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178098.2 NP_835199.1 4 425
GenBank mRNA AK015635.1 - 4 395
AK145933.1 - 4 426
AY146988.1 - 4 425
BC061218.1 - 4 412 30281034

Back To Top

More Information


Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
proteolysis involved in cellular protein catabolic process
cysteine-type endopeptidase activity
cysteine-type peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0