siRNA Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175006.2 | NP_778171.2 | 8 | 1989 | ||
| GenBank mRNA | AK083772.1 | - | 8 | 1989 | ||
| BC138151.1 | - | 8 | 1911 | 8861170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou6f2| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175006.2 | NP_778171.2 | 8 | 1989 | ||
| GenBank mRNA | AK083772.1 | - | 8 | 1989 | ||
| BC138151.1 | - | 8 | 1911 | 8861170 | ||