This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TYRO3 SIRNA Assays ›
Gene Symbol:
Tyro3
Gene Name:
TYRO3 protein tyrosine kinase 3
Gene Aliases:
AI323366, Brt, Dtk, Etk-2, Rse, Sky, TK19-2, Tif, etk2/tyro3
Chromosome Location:
Chr.2: 119797740 - 119818104 on Build GRCm38
UniGene ID:
Mm.2901
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS212193
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tyro3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290800.1 NP_001277729.1 3, 4 1118
NM_019392.2 NP_062265.2 3, 4 654
GenBank mRNA AB000828.1 - 3, 4 391
AK141198.1 - 3, 4 425
BC066058.1 - 3, 4 654 6834015
BC082325.1 - 3, 4 602 30356080
D17393.1 - 3, 4 1099
U05683.1 - 3, 4 439
U18342.1 - 3, 4 571
U18343.1 - 3, 4 1114
U18933.1 - 3, 4 590
X78103.1 - 3, 4 598

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

natural killer cell differentiation
protein phosphorylation
cell adhesion
spermatogenesis
phosphorylation
peptidyl-tyrosine phosphorylation
forebrain cell migration
platelet activation
secretion by cell
negative regulation of toll-like receptor signaling pathway
substrate adhesion-dependent cell spreading
ovulation cycle
apoptotic cell clearance
protein kinase B signaling
negative regulation of neuron apoptotic process
negative regulation of innate immune response
protein autophosphorylation
negative regulation of inflammatory response
negative regulation of lymphocyte activation
vagina development
neuron cellular homeostasis
platelet aggregation
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phosphatidylinositol 3-kinase binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0