siRNA Details
| Gene Symbol: Cbwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146097.3 | NP_666209.1 | 14, 15 | 1154 | ||
| GenBank mRNA | AK076027.1 | - | 14, 15 | 1108 | ||
| AK132291.1 | - | 14, 15 | 1140 | |||
| BC018472.1 | - | 14, 15 | 1096 | 3661842 | ||
| BC027643.1 | - | 3, 4 | 245 | |||
| BC053745.1 | - | 14, 15 | 1189 | 6505520 | ||

Cbwd1
Genomic Map