siRNA Details
| Gene Symbol: Btbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025431.1 | NP_001020602.1 | 5 | 1453 | ||
| NM_145534.2 | NP_663509.2 | 4 | 1755 | |||
| GenBank mRNA | AB093275.1 | - | 4 | 1755 | ||
| AK052530.1 | - | 5 | 1414 | |||
| AK161924.1 | - | 5 | 1410 | |||
| BC018174.1 | - | 4 | 1732 | 4011475 | ||
| BC062968.1 | - | 4 | 1451 | 6408511 | ||

Btbd3
Genomic Map