siRNA Details
| Gene Symbol: Haus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173400.2 | NP_775576.2 | 10 | 1369 | ||
| GenBank mRNA | AK036635.1 | - | 1 | 34 | ||
| AK051016.1 | - | 10 | 1390 | |||
| AK077681.1 | - | 10 | 1391 | |||
| AK136163.1 | - | 1 | 89 | |||
| AK169222.1 | - | 10 | 1371 | |||
| AK173206.1 | - | 10 | 1341 | |||
| BC064725.1 | - | 5 | 566 | |||
| BC068237.1 | - | 10 | 1369 | 30550558 | ||

Haus6
Genomic Map