siRNA Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172216.1 | NP_001165687.1 | 4 | 739 | ||
| NM_172727.3 | NP_766315.1 | 4 | 739 | |||
| GenBank mRNA | AK052323.1 | - | 4 | 653 | ||
| AK140239.1 | - | 4 | 645 | |||
| BC039996.1 | - | 1 | 66 | |||
| BC055818.1 | - | 4 | 667 | 4487036 | ||

Fam221a
Genomic Map