siRNA Details
| Gene Symbol: Ankrd26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081112.1 | NP_001074581.1 | 25 | 3797 | ||
| GenBank mRNA | BC008111.1 | - | 5 | 1343 | ||
| BC056364.1 | - | 8 | 1856 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd26| Gene Symbol: Ankrd26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081112.1 | NP_001074581.1 | 25 | 3797 | ||
| GenBank mRNA | BC008111.1 | - | 5 | 1343 | ||
| BC056364.1 | - | 8 | 1856 | |||