siRNA Details
| Gene Symbol: Rsf1os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037770.1 | - | 462 | |||
| Gene Symbol: Rsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081267.2 | NP_001074736.1 | 13 | 3281 | ||
| GenBank mRNA | BC054454.1 | - | 8 | 771 | ||
| BC085127.1 | - | 12 | 4210 | |||

Rsf1os2
Genomic Map