siRNA Details
| Gene Symbol: Brip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178309.2 | NP_840094.1 | 10 | 1537 | ||
| GenBank mRNA | AK033205.1 | - | 10 | 1508 | ||
| AK051878.1 | - | 10 | 1538 | |||
| AK145709.1 | - | 10 | 1538 | |||
| AK169440.1 | - | 10 | 1484 | |||
| AK169823.1 | - | 9 | 1274 | |||
| BC094252.1 | - | 10 | 1486 | 6401109 | ||

Brip1
Genomic Map