This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other INSRR SIRNA Assays ›
Gene Symbol:
Insrr
Gene Name:
insulin receptor-related receptor
Gene Aliases:
Irr
Chromosome Location:
Chr.3: 87796908 - 87816101 on Build GRCm38
UniGene ID:
Mm.42041
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS215774
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Insrr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011832.2 NP_035962.2 14 3174
GenBank mRNA AB007135.1 - 14 3063
BC137855.1 - 14 3031 8860874

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
phosphorylation
peptidyl-tyrosine phosphorylation
male sex determination
actin cytoskeleton reorganization
protein autophosphorylation
cellular response to alkaline pH
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phosphatidylinositol 3-kinase binding
insulin receptor substrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline