siRNA Details
| Gene Symbol: Gpr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173365.2 | NP_775541.1 | 2 | 598 | ||
| GenBank mRNA | AK040547.1 | - | 2 | 599 | ||
| AY255575.1 | - | 1 | 143 | |||
| BC117783.2 | - | 2 | 460 | 40092101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr20| Gene Symbol: Gpr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173365.2 | NP_775541.1 | 2 | 598 | ||
| GenBank mRNA | AK040547.1 | - | 2 | 599 | ||
| AY255575.1 | - | 1 | 143 | |||
| BC117783.2 | - | 2 | 460 | 40092101 | ||