siRNA Details
| Gene Symbol: Olfr198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011808.1 | NP_001011808.1 | 1 | 214 | ||
| GenBank mRNA | BC147215.1 | - | 1 | 309 | 9056251 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr198| Gene Symbol: Olfr198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011808.1 | NP_001011808.1 | 1 | 214 | ||
| GenBank mRNA | BC147215.1 | - | 1 | 309 | 9056251 | |