This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR427 SIRNA Assays ›
Gene Symbol:
Olfr427 Olfr426
Gene Name:
olfactory receptor 427
olfactory receptor 426
Gene Aliases:
MOR105-7, MOR105-9
GA_x6K02T2P20D-21063569-21062619, MOR105-13_p, MOR105-8
Chromosome Location:
Chr.1: 174099460 - 174100407 on Build GRCm38
Chromosome Location:
-
UniGene ID:
Mm.377354
Mm.489574
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID MSS217558
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr427
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207158.1 NP_997041.1 1 817
GenBank mRNA BC104103.2 - 1 817 40043136
BC104104.2 - 1 817 40043137
BC106807.2 - 1 817 40042912
BC106808.2 - 1 817 40042917
Gene Symbol: Olfr426
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206926.1 NP_001193855.1 1 817

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline