This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GCDH SIRNA Assays ›
Gene Symbol:
Gcdh
Gene Name:
glutaryl-Coenzyme A dehydrogenase
Gene Aliases:
9030411L18, AI266902, D17825, GCD
Chromosome Location:
Chr.8: 84886387 - 84893921 on Build GRCm38
UniGene ID:
Mm.2475
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS219218
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gcdh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044744.1 NP_001038209.2 8 1032
NM_008097.2 NP_032123.3 7 1089
GenBank mRNA AK078866.1 - 5 642
AK079500.1 - 1 140
AK165406.1 - 7 915
BC061158.1 - 7 919 30314273
U18992.1 - 7 870

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

acyl-CoA metabolic process
metabolic process
fatty acid oxidation
fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
glutaryl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-v5kch:80/100.66.78.106:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0