This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADAMTS16 SIRNA Assays ›
Gene Symbol:
Adamts16
Gene Name:
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 16
Gene Aliases:
-
Chromosome Location:
Chr.13: 70727798 - 70841848 on Build GRCm38
UniGene ID:
Mm.115970
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS219363
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adamts16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172053.2 NP_742050.2 3 657
GenBank mRNA AK031314.1 - 3 623
AK134523.1 - 3 643
AK173338.1 - 3 593
BC034739.1 - 3 670 4951547

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003748684.

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
regulation of systemic arterial blood pressure
proteolysis
regulation of cilium assembly
molecular_function
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline