siRNA Details
| Gene Symbol: Serpinb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173052.2 | NP_766640.1 | 2 | 209 | ||
| GenBank mRNA | AF426025.1 | - | 1 | 92 | ||
| AK018226.1 | - | 2 | 188 | |||
| BC030354.1 | - | 2 | 154 | 5367365 | ||
| BY109795.1 | - | 2 | 209 | |||

Serpinb1b
Genomic Map