This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other A230046K03RIK SIRNA Assays ›
Gene Symbol:
A230046K03Rik
Gene Name:
RIKEN cDNA A230046K03 gene
Gene Aliases:
AA589518, Kiaa1033, mKIAA1033
Chromosome Location:
Chr.10: 83543941 - 83596486 on Build GRCm38
UniGene ID:
Mm.491194
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS220076
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A230046K03Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033375.2 NP_001028547.2 10 868
GenBank mRNA AK038582.1 - 10 826
AK043911.1 - 10 839
AK129269.1 - 5 398
AK145000.1 - 10 850
AK166233.1 - 10 841
BC020314.2 - 7 503
BC145215.1 - 10 1063

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
endosome organization
protein transport
endosomal transport
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-r9tfj:80/100.66.76.159:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline