siRNA Details
| Gene Symbol: Phf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113354.1 | NP_001106825.1 | 3 | 981 | ||
| NM_177201.5 | NP_796175.1 | 3 | 981 | |||
| GenBank mRNA | AK036609.1 | - | 3 | 355 | ||
| AK040943.1 | - | 3 | 350 | |||
| AK040969.1 | - | 3 | 982 | |||
| AK122447.1 | - | 2 | 72 | |||
| AK134137.1 | - | 3 | 318 | |||
| AK147939.1 | - | 3 | 368 | |||
| BC138899.1 | - | 3 | 250 | 8861921 | ||

Phf8
Genomic Map