siRNA Details
| Gene Symbol: Ppip5k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178795.4 | NP_848910.3 | 6 | 681 | ||
| GenBank mRNA | AF502585.1 | - | 6 | 683 | ||
| AK046696.1 | - | 7 | 723 | |||
| AK078268.1 | - | 7 | 718 | |||
| AK083140.1 | - | 6 | 692 | |||
| AK122264.1 | - | 6 | 647 | |||
| AK160847.1 | - | 5 | 1253 | |||
| AK162596.1 | - | 7 | 725 | |||
| BC065138.1 | - | 6 | 684 | |||
| BC138351.1 | - | 5 | 579 | 8861371 | ||

Ppip5k1
Genomic Map