siRNA Details
| Gene Symbol: 4933402J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177901.3 | NP_808569.1 | 6 | 876 | ||
| GenBank mRNA | AK077084.1 | - | 6 | 877 | ||
| AK132946.1 | - | 6 | 932 | |||
| BC117078.1 | - | 6 | 869 | 40126397 | ||
| BC120779.1 | - | 6 | 869 | 40129702 | ||

4933402J07Rik
Genomic Map