siRNA Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177390.3 | NP_796364.2 | 13, 14 | 1839 | ||
| GenBank mRNA | AK049374.1 | - | 13, 14 | 1840 | ||
| BC039700.1 | - | 8, 9 | 971 | |||
| BC150765.1 | - | 13, 14 | 1705 | 9087676 | ||

Myo1d
Genomic Map